missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ trans-5-Hydroxyferulic acid, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 110642-42-7 |
| Product Line | phyproof |
| Linear Formula | C10H10O5 |
| MDL Number | MFCD03002791 |
| Synonym | (2 E)-3-(3,4-Dihydroxy-5-methoxyphenyl)-2-propenoic acid |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 210.18 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG