missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Tiliroside, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 20316-62-5 |
| Product Line | phyproof |
| Linear Formula | C30H26O13 |
| MDL Number | MFCD00017454 |
| Synonym | Astragalin 6 ′′-trans-p-coumarate; Kaempferol 3-O-beta-D-(6 ′′-O-(E)-p-coumaroyl)glucopyranoside; Potengriffioside A |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 594.52 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG