missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Sophoridine, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 6882-68-4 |
| Product Line | phyproof |
| Linear Formula | C15H24N2 O |
| MDL Number | MFCD03850811 |
| Synonym | 5 beta-Matridin 15-one; 5-Epidihydrosophocarpine; Dihydro 5-episophocarpine |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 248.36 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG