missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Silybin B, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 142797-34-0 |
| Product Line | phyproof |
| Linear Formula | C25H22O10 |
| MDL Number | MFCD18827498 |
| Synonym | (2 R,3 R)-3,5,7-Trihydroxy-2-[(2 S,3 S)-3-(4-hydroxy-3-methoxyphenyl)-2-hydroxymethyl-2,3-dihydrobenzo[1,4 ]dioxin-6-yl]-4-chromanone; (2 S,3 S)-2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-6-[(2 R,3 R)-3,5,7-trihydroxy-4-oxobenzopyran-2-yl]benzodioxin |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 482.44 |
| Percent Purity | ≥98% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG