missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Sesamolin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 526-07-8 |
| Product Line | phyproof |
| Linear Formula | C20H18O7 |
| Synonym | 5-[(1 S,3 aR,4 R,6 aR)-4-(1,3-Benzodioxol-5-yloxy)tetrahydro-1 H,3 H-furo[3,4-c]furan-1-yl]-1,3-benzodioxole |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 370.35 |
| Percent Purity | ≥95% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG