missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Sennoside C, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002131
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002131 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002131 Supplier Merck Emd Millipore Supplier No. PHL8053410MG
Only null left

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 37271-16-2
Product Line phyproof
Flash Point 344.4°C
Linear Formula C42H40O19
Type Reference Substance
Quantity 10 mg
Formula Weight 848.76
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG