missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ α-Santonin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002071
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002071 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002071 Supplier Merck Emd Millipore Supplier No. PHL80931100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 481-06-1
Product Line phyproof
Linear Formula C15H18O3
MDL Number MFCD00135865
Melting Point 172°C to 173°C
Synonym (-)-alpha-Santonin; (3 S,5 AS,9 bS)-3 A,5,5 A,9 b-Tetrahydro-3,5 A,9-trimethylnaphtho[1,2-b]furan-2,8(3 H,4 H)dione; Semenen
Type Reference Substance
Quantity 100 mg
Formula Weight 246.30
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG