missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Sakuranetin, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 520-29-6 |
| Product Line | phyproof |
| Purity Grade Notes | For this product we use the non-stereospecific CAS number. It is expected that the natural enantiomer is predominant, however the substance is prone to racemization in solution. The product is therefore not specified stereospecifically and is only recommended to be used for non-stereospecific analysis. |
| Linear Formula | C16H14O5 |
| MDL Number | MFCD00075651 |
| Synonym | 4′-5-Dihydroxy-7-methoxyflavanone; 7-O-Methylnaringenin; Naringenin 7-O-methyl ether |
| Type | Reference Substance |
| Quantity | 20 mg |
| Formula Weight | 286.28 |
| Percent Purity | ≥98% (HPLC) |
| Show More |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG