missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Robinetin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 490-31-3 |
| Product Line | phyproof |
| Linear Formula | C15H10O7 |
| MDL Number | MFCD00016783 |
| Synonym | 3,3′,4′,5′,7-Pentahydroxyflavone; 3,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)-4 H-1-benzopyran-4-one; Norkanugin |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 302.24 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG