missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Pyrogallol, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001848
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001848 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001848 Supplier Merck Emd Millipore Supplier No. PHL89772100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 87-66-1
Product Line phyproof
Vapor Pressure 10 mmHg (167.7°C)
Vapor Density 4.4 (vs. air)
Linear Formula C6H3(OH)3
MDL Number MFCD00002192
Melting Point 133°C to 134°C
Boiling Point 309°C
Synonym 1,2,3-Trihydroxybenzene; 2,3-Dihydroxyphenol; Pyrogallic acid
Type Reference Substance
Quantity 100 mg
Formula Weight 126.11
Percent Purity ≥98% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG