missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Puerarin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002134
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002134 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002134 Supplier Merck Emd Millipore Supplier No. PHL8931810MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 3681-99-0
Product Line phyproof
Linear Formula C21H20O9
MDL Number MFCD00076007
Synonym 8-(beta-D-Glucopyranosyl-7-hydroxy-3- (4-hydroxyphenyl)-4 H-1-benzopyran-4-one; Kakonein
Type Reference Substance
Quantity 10 mg
Formula Weight 416.38
Percent Purity ≥95% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG