missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Psoralidin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 18642-23-4 |
| Product Line | phyproof |
| Linear Formula | C20H16O5 |
| MDL Number | MFCD10566617 |
| Synonym | 3,9-Dihydroxy-2-(3-methyl-2-butenyl)-6 H-benzofuro[3,2-c][1 ]benzopyran-6-one; 3,9-Dihydroxy-2-prenylcoumestan |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 336.34 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG