missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Pseudolaric acid C, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001867
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001867 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001867 Supplier Merck Emd Millipore Supplier No. PHL8263510MG
Only null left

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 82601-41-0
Product Line phyproof
Linear Formula C21H26O7
MDL Number MFCD16660657
Type Reference Substance
Quantity 10 mg
Formula Weight 390.43
Percent Purity ≥98% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG