missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Pseudolaric Acid B, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001869
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001869 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001869 Supplier Merck Emd Millipore Supplier No. PHL8052310MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 82508-31-4
Product Line phyproof
Linear Formula C23H28O8
MDL Number MFCD26960905
Synonym (-)-Pseudolaric acid B; O-Acetylpseudolaric acid C; Pseudolarix acid B
Type Reference Substance
Quantity 10 mg
Formula Weight 432.46
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG