missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Protopine, ≥98% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 130-86-9 |
| Product Line | phyproof |
| Linear Formula | C20H19 NO5 |
| MDL Number | MFCD00022322 |
| Synonym | 4,6,7,14-Tetrahydro-5-methylbis[1,3 ]benzodioxolo[4,5-c:5′,6′-g]azecin-13(5 H)-one |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 353.37 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG