missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Poncirin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 14941-08-3 |
| Product Line | phyproof |
| Linear Formula | C28H34O14 |
| MDL Number | MFCD00017476 |
| Synonym | -5,7-Dihydroxy-4′-methoxyflavanone-7-[2-O-(alpha-L-rhamnopyranosyl)-beta-D-glucopyranoside]; 4′-O-Methylnaringin; Isosakuranetin-7-neohesperidoside |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 594.56 |
| Percent Purity | ≥95% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG