missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Pinosylvin monomethyl ether, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002146
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002146 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002146 Supplier Merck Emd Millipore Supplier No. PHL8051710MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 35302-70-6
Product Line phyproof
Linear Formula C15H14O2
MDL Number MFCD00210546
Synonym (E)-3-Hydroxy-5-methoxystilbene; (E)-3-Methoxy-5-(2-phenylethenyl)phenol; 5-Methoxy-3-stilbenol; Pinosylvin 3-methyl ether
Type Reference Substance
Quantity 10 mg
Formula Weight 226.27
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG