missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ (+)-Pinoresinol, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | (+)-Pinoresinol |
| CAS | 487-36-5 |
| Product Line | phyproof |
| Linear Formula | C20H22O6 |
| MDL Number | MFCD07784516 |
| Synonym | Pinoresinol; (+)-Pinoresinol; 4,4′-((1 S,3 aR,4 S,6 aR)-Hexahydrofuro[3,4-c]furan-1,4-diyl)bis(2-methoxyphenol); 4,4′-[(1 S,3 aR,4 S,6 aR)-Tetrahydro-1 H,3 H-furo[3,4-c]furan-1,4-diyl]bis(2-methoxyphenol) |
| Type | Reference Substance |
| Industry Type | Laboratory Use |
| Molecular Weight (g/mol) | 358.39 |
| Quantity | 10 mg |
| Show More |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG