missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Periplocin, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 13137-64-9 |
| Product Line | phyproof |
| Linear Formula | C36H56O13 |
| MDL Number | MFCD01745141 |
| Synonym | Periplocoside |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 696.82 |
| Percent Purity | ≥98% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG