missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Peonidin 3-glucoside chloride, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001918
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001918 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001918 Supplier Merck Emd Millipore Supplier No. PHL897545MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 6906-39-4
Product Line phyproof
Linear Formula C22H23 ClO11
MDL Number MFCD00189435
Synonym Peonidin 3-O-glucoside chloride; 3-(Glucosyloxy)-4′,5,7-trihydroxy-3′-methoxyflavylium chloride; Glucopeonidin chloride
Type Reference Substance
Quantity 5 mg
Formula Weight 498.86
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG