missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Parishin E, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 952068-57-4 |
| Product Line | phyproof |
| Linear Formula | C19H24O13 |
| MDL Number | MFCD29048569 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 460.39 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG