missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Obtusifolin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002081
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002081 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002081 Supplier Merck Emd Millipore Supplier No. PHL8388010MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 477-85-0
Product Line phyproof
Linear Formula C16H12O5
MDL Number MFCD18452296
Synonym 2,8-Dihydroxy-1-methoxy-3-methyl-9,10-anthracenedione
Type Reference Substance
Quantity 10 mg
Formula Weight 284.26
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG