missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Neohesperidin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 13241-33-3 |
| Product Line | phyproof |
| Linear Formula | C28H34O15 |
| MDL Number | MFCD16661259 |
| Synonym | -4′-Methoxy-3′,5,7-trihydroxyflavanone-7-[2-O-(alpha-L-rhamnopyranosyl)-beta-D-glucopyranoside]; Hesperetin 7-O-neohesperidoside; NSC 31048 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 610.56 |
| Percent Purity | ≥95% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG