missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Naringenin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001930
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001930 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001930 Supplier Merck Emd Millipore Supplier No. PHL8973825MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 67604-48-2
Product Line phyproof
Purity Grade Notes For this product we use the non-stereospecific CAS number. It is expected that the natural enantiomer is predominant, however the substance is prone to racemization in solution. The product is therefore not specified stereospecifically and is only recommended to be used for non-stereospecific analysis.
Linear Formula C15H12O5
MDL Number MFCD00006844
Synonym 4′,5,7-Trihydroxyflavanone
Type Reference Substance
Quantity 25 mg
Formula Weight 272.25
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG