missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ N-Vanillylnonanamide, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002225
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002225 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002225 Supplier Merck Emd Millipore Supplier No. PHL8982025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 2444-46-4
Product Line phyproof
Linear Formula C17H27 NO3
MDL Number MFCD00017286
Synonym Capsaicin synthetic; N-(4-Hydroxy-3-methoxybenzyl)nonanamide; N-Vanillylpelargonamide; Nonanoic acid vanillylamide; Nonylvanylamide; Pelargonic acid vanillylamide; Pseudocapsaicin
Type Reference Substance
Quantity 25 mg
Formula Weight 293.40
Percent Purity ≥95% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG