missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Mollugin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001996
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001996 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001996 Supplier Merck Emd Millipore Supplier No. PHL8049910MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 55481-88-4
Product Line phyproof
Linear Formula C17H16O4
MDL Number MFCD02752331
Synonym 6-Hydroxy-2,2-dimethyl-2 H-naphtho(1,2-b)pyran-5-carboxylic acid methyl ester; Rubimaillin
Type Reference Substance
Quantity 10 mg
Formula Weight 284.31
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG