missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Methyl gallate, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001804
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001804 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001804 Supplier Merck Emd Millipore Supplier No. PHL82592100MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 99-24-1
Product Line phyproof
Linear Formula (HO)3C6H2 CO2 CH3
MDL Number MFCD00002194
Synonym Gallic acid methyl ester; Methyl 3,4,5-trihydroxybenzoate
Type Reference Substance
Quantity 100 mg
Formula Weight 184.15
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG