missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Manassantin A, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 88497-87-4 |
| Product Line | phyproof |
| Linear Formula | C42H52O11 |
| Synonym | Saucernetin A |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 732.86 |
| Percent Purity | ≥98% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG