missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Magnolin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 31008-18-1 |
| Product Line | phyproof |
| Linear Formula | C23H28O7 |
| MDL Number | MFCD07783722 |
| Synonym | (1 S,3 aR,4 S,6 aR)-1-(3,4-Dimethoxyphenyl)tetrahydro-4-(3,4,5-trimethoxyphenyl)-1 H,3 H-furo[3,4-c]furan; Medioresinol dimethyl ether |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 416.46 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG