missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Lutein, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002367
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002367 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002367 Supplier Merck Emd Millipore Supplier No. PHL897235MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 127-40-2
Product Line phyproof
Linear Formula C40H56O2
MDL Number MFCD08435941
Melting Point 195°C
Synonym Xanthophyll; alpha-Carotene-3,3′-diol; beta, epsilon-carotene; beta, epsilon-carotene-3,3′-diol; Lutein
Type Reference Substance
Quantity 5 mg
Formula Weight 568.87
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG