missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Linalyl acetate, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111002395
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002395 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002395 Supplier Merck Emd Millipore Supplier No. PHL89243100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 115-95-7
Product Line phyproof
Vapor Pressure 0.1 mmHg (20°C)
Vapor Density 6.8 (vs. air)
Density 0.901 g/mL (at 25°C)
Flash Point 85°C
Linear Formula CH3 CO2 C(CH=CH2)(CH3)CH2 CH2 CH=C(CH3)2
MDL Number MFCD00008907
Boiling Point 220°C
Synonym 3,7-Dimethyl-1,6-octadien-3-yl acetate; Bergamol
Type Reference Substance
Quantity 100 mg
Formula Weight 196.29
Percent Purity ≥90% (GC)
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG