missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Limonin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 1180-71-8 |
| Product Line | phyproof |
| Linear Formula | C26H30O8 |
| MDL Number | MFCD00075922 |
| Synonym | Citrolimonin; Dictamnolactone; Evodin; Limonoic acid 3,19:16,17 dilactone; Obaculactone |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 470.51 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG