missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Lavandulol, ≥90% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 58461-27-1 |
| Product Line | phyproof |
| Flash Point | 98°C |
| Linear Formula | C10H18 O |
| MDL Number | MFCD00674532 |
| Synonym | 2-Isopropenyl-5-methyl-4-hexen-1-ol |
| Type | Reference Substance |
| Quantity | 100 mg |
| Formula Weight | 154.25 |
| Percent Purity | ≥90% (GC) |
| Show More |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG