missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Kaempferol 3-glucoside, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002076
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002076 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002076 Supplier Merck Emd Millipore Supplier No. PHL8923710MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 480-10-4
Product Line phyproof
Linear Formula C21H20O11
MDL Number MFCD00075932
Synonym Kaempferol 3-beta-D-glucopyranoside; 3,4′,5,7-Tetrahydroxyflavone 3-glucoside; 3-(beta-D-Glucopyranosyloxy)-5,7-dihydroxy-2-(4-methoxyphenyl)-4 H-1-benzopyran-4-one; 3-Glucosylkaempferol; Astragalin; Kaempferol 3-glucoside
Type Reference Substance
Quantity 10 mg
Formula Weight 448.38
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG