missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Hesperetin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001917
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001917 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001917 Supplier Merck Emd Millipore Supplier No. PHL8922220MG
Only null left

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 69097-99-0
Product Line phyproof
Purity Grade Notes For this product we use the non-stereospecific CAS number. It is expected that the natural enantiomer is predominant, however the substance is prone to racemization in solution. The product is therefore not specified stereospecifically and is only recommended to be used for non-stereospecific analysis.
Linear Formula C16H14O6
MDL Number MFCD00016956
Synonym 3′,5,7-Trihydroxy-4′-methoxyflavanone; 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-chromanone
Type Reference Substance
Quantity 20 mg
Formula Weight 302.28
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG