missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Helicid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001879
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001879 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001879 Supplier Merck Emd Millipore Supplier No. PHL8255625MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 80154-34-3
Product Line phyproof
Linear Formula C13H16O7
MDL Number MFCD00210992
Synonym 4-(beta-D-Allopyranosyloxy)benzaldehyde
Type Reference Substance
Quantity 25 mg
Formula Weight 284.26
Percent Purity ≥98% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG