missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Hecogenin, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111002089
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002089 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002089 Supplier Merck Emd Millipore Supplier No. PHL8350810MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 467-55-0
Product Line phyproof
Linear Formula C27H42O4
MDL Number MFCD00067285
Synonym 12-Oxotigogenin; 3 beta-Hydroxy-5 alpha-spirostan-12-one; 5 alpha-Spirostan-3 beta-ol-12-one; Gekogenin; Hocogenin
Type Reference Substance
Quantity 10 mg
Formula Weight 430.62
Percent Purity ≥90% (GC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG