missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Gracillin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002271
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002271 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002271 Supplier Merck Emd Millipore Supplier No. PHL8350310MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 19083-00-2
Product Line phyproof
Linear Formula C45H72O17
Synonym Diosgenin 3-gracillimatrioside; Dulcamarin C; Lilioglycoside G
Type Reference Substance
Quantity 10 mg
Formula Weight 885.04
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG