missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Glucoraphenin potassium salt, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 108844-81-1 |
| Product Line | phyproof |
| Linear Formula | C12H20 KNO10S3 |
| Synonym | 4-Methylsulfinyl 3-butenylglucosinolate potassium salt |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 473.58 |
| Percent Purity | ≥90% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG