missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ β-Glucogallin, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002351
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002351 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002351 Supplier Merck Emd Millipore Supplier No. PHL8325010MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 13405-60-2
Product Line phyproof
Linear Formula C13H16O10
MDL Number MFCD11975493
Synonym 1-O-Galloyl-beta-D-glucose; beta-D-Glucopyranose 1-(3,4,5-trihydroxybenzoate); beta-Glucogallin; 1-O-Galloyl-beta-D-glucopyranose
Type Reference Substance
Quantity 10 mg
Formula Weight 332.26
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG