missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Ginsenoside Rh1, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 63223-86-9 |
| Product Line | phyproof |
| Linear Formula | C36H62O9 |
| MDL Number | MFCD09951797 |
| Synonym | (3 beta,6 alpha,12 beta)-3,12,20-Trihydroxydammar-24-en-6-yl-beta-D-glucopyranoside; Prosapogenin A2; Sanchinoside B2; Sanchinoside Rh1 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 638.87 |
| Percent Purity | ≥90% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG