missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Ginsenoside Rd, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002014
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002014 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002014 Supplier Merck Emd Millipore Supplier No. PHL8921110MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 52705-93-8
Product Line phyproof
Linear Formula C48H82O18
Synonym (3 beta,12 beta)-20-(beta-D-Glucopyranosyloxy)-12-hydroxydammar-24-en-3-yl 2-O-beta-D-glucopyranosyl-beta-D-glucopyranoside; Gypenoside VIII
Type Reference Substance
Quantity 10 mg
Formula Weight 947.15
Percent Purity ≥90% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG