missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Garcinone D, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002419
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002419 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002419 Supplier Merck Emd Millipore Supplier No. PHL8254710MG
Only null left

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 107390-08-9
Product Line phyproof
Linear Formula C24H28O7
MDL Number MFCD11111286
Synonym 1,3,6-Trihydroxy-8-(3-hydroxy-3-methylbutyl)-7-methoxy-2-(3-methyl-2-buten-1-yl)-9 H-xanthen-9-one
Type Reference Substance
Quantity 10 mg
Formula Weight 428.47
Percent Purity ≥95% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG