missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Galantamine hydrobromide, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002266
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002266 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002266 Supplier Merck Emd Millipore Supplier No. PHL8966650MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 4-4-1953
Product Line phyproof
Linear Formula C17H21 NO3 · HBr
MDL Number MFCD26142877
Synonym Galanthamine hydrobromide
Type Reference Substance
Quantity 50 mg
Formula Weight 368.27
Percent Purity ≥95% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG