missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Frangulin B, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002332
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002332 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002332 Supplier Merck Emd Millipore Supplier No. PHL8919610MG

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 14101-04-3
Product Line phyproof
Linear Formula C20H18O9
MDL Number MFCD00016362
Synonym 3-(D-Apio-beta-D-furanosyloxy)-1,8-dihydroxy-6-methyl-9,10-anthracenedione
Type Reference Substance
Quantity 10 mg
Formula Weight 402.35
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG