missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Eriodictyol, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002113
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002113 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002113 Supplier Merck Emd Millipore Supplier No. PHL8044420MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 4049-38-1
Product Line phyproof
Purity Grade Notes For this product we use the non-stereospecific CAS number. It is expected that the natural enantiomer is predominant, however the substance is prone to racemization in solution. The product is therefore not specified stereospecifically and is only recommended to be used for non-stereospecific analysis.
Linear Formula C15H12O6
MDL Number MFCD00017312
Synonym 3·,4·,5,7-Tetrahydroxyflavanone
Type Reference Substance
Quantity 20 mg
Formula Weight 288.25
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG