missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Epoxybergamottin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 206978-14-5 |
| Product Line | phyproof |
| Linear Formula | C21H22O5 |
| MDL Number | MFCD16251690 |
| Synonym | 6′,7′-Epoxybergamottin; 4-[[(2 E)-5-(3,3-Dimethyl-2-oxiranyl)-3-methyl-2-penten-1-yl]oxy]-7 H-furo[3,2-g][1 ]benzopyran-7-one; 5-(6′,7′-Epoxy)geranyloxypsoralen; Bergamottin 6′,7′-epoxide; Epoxybergamottin |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 354.40 |
| Percent Purity | ≥95% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG