missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Dipsacoside B, ≥95% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 33289-85-9 |
| Product Line | phyproof |
| Linear Formula | C53H86O22 |
| MDL Number | MFCD03424249 |
| Synonym | Glycoside L-G3; Tauroside G3; Tauroside St-G3 |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 1075.24 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG