missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Dihydrocapsaicin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 19408-84-5 |
| Product Line | phyproof |
| Linear Formula | C18H29 NO3 |
| MDL Number | MFCD00057794 |
| Synonym | 6,7-Dihydrocapsaicin; 8-Methyl-N-vanillylnonanamide; N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methylnonenamide |
| Type | Reference Substance |
| Quantity | 20 mg |
| Formula Weight | 307.43 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG