missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Cyanidin chloride, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 528-58-5 |
| Product Line | phyproof |
| Linear Formula | C15H11 ClO6 |
| MDL Number | MFCD00017582 |
| Synonym | 3,3′,4,5,7-Pentahydroxyflavylium chloride; Cyanidol chloride |
| Type | Reference Substance |
| Quantity | 20 mg |
| Formula Weight | 322.70 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG